siRNA Details
Gene Symbol: Serpinb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173052.2 | NP_766640.1 | 7 | 1286 | ||
GenBank mRNA | AK018226.1 | - | 7 | 1261 | ||
BC030354.1 | - | 7 | 1231 | 5367365 |

More Information
Other Species Targeted

Gene Ontology Categories:
