siRNA Details
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081278.2 | NP_001074747.2 | 1, 2 | 1237 | ||
GenBank mRNA | AK031120.1 | - | 1, 2 | 1081 | ||
AK044719.1 | - | 1, 2 | 879 | |||
BC117869.1 | - | 1, 2 | 713 | 40095526 | ||
BC117870.1 | - | 1, 2 | 713 | 40095531 |

More Information
Other Species Targeted

Gene Ontology Categories:
