siRNA Details
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178246.3 | NP_839977.2 | 3 | 409 | ||
GenBank mRNA | AK129286.1 | - | 3 | 388 | ||
AK143911.1 | - | 3 | 410 | |||
AK155787.1 | - | 3 | 398 | |||
AK172217.1 | - | 2 | 287 | |||
BC024683.1 | - | 3 | 361 | 3992000 | ||
BC100408.1 | - | 3 | 358 | 30734268 |

More Information
Other Species Targeted

Gene Ontology Categories:
