siRNA Details
Gene Symbol: Cypt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173367.2 | NP_775543.2 | 2 | 616 | ||
GenBank mRNA | AF463500.1 | - | 2 | 596 | ||
AK005674.1 | - | 2 | 629 | |||
AV047945.2 | - | 1 | 279 | |||
BC048527.1 | - | 2 | 616 | 6772990 |
