siRNA Details
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081278.2 | NP_001074747.2 | 2 | 1291 | ||
GenBank mRNA | AK031120.1 | - | 2 | 1135 | ||
AK039337.1 | - | 2 | 1292 | |||
AK044719.1 | - | 2 | 933 | |||
BC064433.1 | - | 2 | 915 | |||
BC080762.1 | - | 2 | 909 | |||
BC117869.1 | - | 2 | 767 | 40095526 | ||
BC117870.1 | - | 2 | 767 | 40095531 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
