siRNA Details
Gene Symbol: ERC2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024615.1 | 1 | 712 | |||
GenBank mRNA | BC069453.1 | - | 1 | 374 | ||
BC074933.2 | - | 1 | 345 | |||
BC074934.2 | - | 1 | 345 | |||
U88965.1 | - | 1 | 624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
