siRNA Details
Gene Symbol: Nbeal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173444.2 | NP_775620.2 | 44, 45 | 7054 | ||
GenBank mRNA | AK041108.1 | - | 4, 5 | 392 | ||
BC055813.1 | - | 7, 8 | 783 | |||
BC060141.1 | - | 20, 21 | 3185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
