siRNA Details
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178246.3 | NP_839977.2 | 10, 11 | 1289 | ||
GenBank mRNA | AK129286.1 | - | 10, 11 | 1268 | ||
AK133688.1 | - | 1, 2 | 113 | |||
AK143911.1 | - | 10, 11 | 1291 | |||
AK155787.1 | - | 10, 11 | 1278 | |||
BC024683.1 | - | 10, 11 | 1241 | 3992000 | ||
BC066787.1 | - | 4, 5 | 516 | |||
BC100408.1 | - | 10, 11 | 1238 | 30734268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
