siRNA Details
Gene Symbol: Tbc1d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081278.2 | NP_001074747.2 | 12 | 3049 | ||
GenBank mRNA | AK039337.1 | - | 14 | 3239 | ||
AK044719.1 | - | 12 | 2691 | |||
AK047705.1 | - | 2 | 220 | |||
AK171689.1 | - | 1 | 240 | |||
AK172575.1 | - | 1 | 120 | |||
AK220347.1 | - | 11 | 1456 | |||
BC042747.1 | - | 8 | 1051 | |||
BC064433.1 | - | 12 | 2673 | |||
BC080762.1 | - | 12 | 2667 | |||
BC117869.1 | - | 12 | 2522 | 40095526 | ||
BC117870.1 | - | 13 | 2549 | 40095531 |

More Information
Other Species Targeted

Gene Ontology Categories:
