siRNA Details
Gene Symbol: Nbeal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173444.2 | NP_775620.2 | 45 | 7089 | ||
GenBank mRNA | AK041108.1 | - | 5 | 427 | ||
BC055813.1 | - | 8 | 818 | |||
BC060141.1 | - | 21 | 3220 |

More Information
Other Species Targeted

Gene Ontology Categories:
