siRNA Details
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029835.1 | NP_084111.1 | 20 | 5019 | ||
GenBank mRNA | AK033104.1 | - | 1 | 565 | ||
BC058634.1 | - | 1 | 231 | |||
BC158103.1 | - | 20 | 4983 | 9088180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
