siRNA Details
Gene Symbol: Nek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293637.1 | NP_001280566.1 | 2 | 685 | ||
NM_001293638.1 | NP_001280567.1 | 2 | 685 | |||
NM_001293639.1 | NP_001280568.1 | 2 | 685 | |||
NM_175089.4 | NP_780298.2 | 2 | 685 | |||
GenBank mRNA | AK045202.1 | - | 2 | 638 | ||
AK077047.1 | - | 3 | 356 | |||
AK173292.1 | - | 2 | 683 | |||
AY850065.1 | - | 2 | 658 | |||
BC070443.1 | - | 2 | 628 | |||
BC157939.1 | - | 2 | 613 | 9088119 | ||
BC158009.1 | - | 2 | 613 | 9007317 | ||
BC172091.1 | - | 2 | 613 | 9054377 |

More Information
Other Species Targeted

Gene Ontology Categories:
