siRNA Details
Gene Symbol: Maml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013813.3 | NP_001013835.2 | 2649 | |||
NM_173776.3 | NP_776137.2 | 4 | 553 | |||
GenBank mRNA | AK031222.1 | - | 4 | 390 | ||
AK129448.1 | - | 4 | 553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
