siRNA Details
Gene Symbol: Nek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293637.1 | NP_001280566.1 | 3 | 744 | ||
NM_001293638.1 | NP_001280567.1 | 3 | 744 | |||
NM_001293639.1 | NP_001280568.1 | 3 | 744 | |||
NM_175089.4 | NP_780298.2 | 3 | 744 | |||
GenBank mRNA | AK077047.1 | - | 4 | 415 | ||
AK173292.1 | - | 3 | 742 | |||
AY850065.1 | - | 3 | 717 | |||
BC070443.1 | - | 3 | 687 | |||
BC157939.1 | - | 3 | 672 | 9088119 | ||
BC158009.1 | - | 3 | 672 | 9007317 | ||
BC172091.1 | - | 3 | 672 | 9054377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
