siRNA Details
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008445.2 | NP_032471.2 | 8 | 4753 | ||
GenBank mRNA | AK050003.1 | - | 1 | 1119 | ||
AK090195.1 | - | 1 | 913 | |||
AK139894.1 | - | 8 | 2429 | |||
AK147572.1 | - | 8 | 4553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
