siRNA Details
Gene Symbol: Vwa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027906.1 | NP_082182.1 | 45 | 6070 | ||
GenBank mRNA | AK030028.1 | - | 9 | 1652 | ||
AK129164.1 | - | 29 | 3989 | |||
BC037705.1 | - | 10 | 1610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
