siRNA Details
Gene Symbol: Srcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051146.1 | - | 5 | 948 | ||
AK142089.1 | - | 5 | 992 | |||
AK154145.1 | - | 6 | 633 | |||
AK169954.1 | - | 5 | 976 | |||
BC085144.1 | - | 5 | 931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
