siRNA Details
Gene Symbol: Mast4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175171.3 | NP_780380.2 | 1, 2 | 650 | ||
GenBank mRNA | AK040801.1 | - | 1, 2 | 611 | ||
AK044801.1 | - | 1, 2 | 619 | |||
AK046702.1 | - | 1, 2 | 622 | |||
AK081750.1 | - | 1, 2 | 607 | |||
AK090136.1 | - | 1, 2 | 622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
