siRNA Details
Gene Symbol: Wdr44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041211.1 | - | 3 | 340 | ||
AK049071.1 | - | 2 | 480 | |||
AK088885.1 | - | 3 | 305 | |||
BB619844.1 | - | 2 | 506 | |||
BC049191.1 | - | 3 | 243 | 6476607 | ||
BC068151.1 | - | 2 | 439 | 30534873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
