siRNA Details
Gene Symbol: Psmb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175204.4 | NP_780413.1 | 1 | 271 | ||
GenBank mRNA | AB299436.1 | - | 1 | 254 | ||
AK017902.1 | - | 1 | 269 | |||
AK037368.1 | - | 1 | 269 | |||
AK041393.1 | - | 1 | 269 | |||
AK162727.1 | - | 1 | 273 | |||
BC117732.1 | - | 1 | 269 | 40089808 | ||
BC117733.1 | - | 1 | 269 | 40089809 |
