siRNA Details
Gene Symbol: Brip1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052488.1 | - | 2 | 635 | ||
AK078546.1 | - | 2 | 668 | |||
AK084670.1 | - | 2 | 662 | |||
AK132401.1 | - | 2 | 668 | |||
BC067002.1 | - | 2 | 677 | 5716200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
