siRNA Details
Gene Symbol: Atp11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293667.1 | NP_001280596.1 | 30 | 6464 | ||
NM_001293668.1 | NP_001280597.1 | 30 | 6445 | |||
NM_015804.3 | NP_056619.1 | 29 | 6360 | |||
GenBank mRNA | AK028779.1 | - | 1 | 1204 | ||
AK078972.1 | - | 1 | 792 | |||
AK147497.1 | - | 30 | 6467 | |||
AK161916.1 | - | 1 | 1387 | |||
BC036322.1 | - | 15 | 4505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
