siRNA Details
Gene Symbol: Zfp935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178875.4 | NP_849206.2 | 3, 4 | 302 | ||
GenBank mRNA | BC047396.1 | - | 3, 4 | 279 | 3979342 | |
BC087936.1 | - | 3, 4 | 302 | |||
BC113193.1 | - | 3, 4 | 298 | 40062446 |
Gene Symbol: 2810408B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013043.1 | - | 720 |
Gene Symbol: AU016916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN455334.1 | - | 3, 4 | 294 |
Gene Symbol: Gm3604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162910.1 | NP_001156382.1 | 3, 4 | 291 | ||
GenBank mRNA | BC049144.1 | - | 3, 4 | 283 |
Gene Symbol: Gm5141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256065.1 | NP_001242994.1 | 3, 4 | 324 | ||
GenBank mRNA | BC064714.1 | - | 3, 4 | 324 |
Gene Symbol: Zfp808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039239.2 | NP_001034328.2 | 3, 4 | 335 | ||
GenBank mRNA | BC061203.1 | - | 3, 4 | 335 | 30248220 |
Gene Symbol: Zfp934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162911.1 | NP_001156383.1 | 3, 4 | 309 | ||
NM_175252.4 | NP_780461.2 | 4, 5 | 513 | |||
GenBank mRNA | AK030776.1 | - | 4, 5 | 516 | ||
BC139791.1 | - | 3, 4 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
