siRNA Details
Gene Symbol: Cflar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289704.2 | NP_001276633.1 | 10 | 4965 | ||
NM_001293804.1 | NP_001280733.1 | 10 | 4948 | |||
NM_001293805.1 | NP_001280734.1 | 8 | 4441 | |||
NM_207653.5 | NP_997536.1 | 10 | 5237 | |||
GenBank mRNA | AK142078.1 | - | 1 | 153 | ||
BC023121.1 | - | 1 | 331 |
Gene Symbol: 1700085D22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054489.1 | - | 1 | 2535 |
Gene Symbol: 4930427A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK220305.1 | - | 4089 |
Gene Symbol: 4933437F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014995.1 | - | 1 | 240 | ||
AK017098.1 | - | 1 | 248 |
Gene Symbol: A230046K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033375.2 | NP_001028547.2 | 33 | 5328 | ||
GenBank mRNA | AK089097.1 | - | 4 | 2135 | ||
AK129269.1 | - | 28 | 4858 | |||
AK161247.1 | - | 10 | 2776 |
Gene Symbol: A430073I11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079808.1 | - | 2 | 794 |
Gene Symbol: A830037D11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080644.1 | - | 1 | 1161 |
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013677.1 | - | 1 | 1015 |
Gene Symbol: Agl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081326.1 | NP_001074795.1 | 34 | 9078 | ||
GenBank mRNA | AK044455.1 | - | 1 | 3053 |
Gene Symbol: Ankfy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009671.5 | NP_033801.4 | 25 | 7836 | ||
GenBank mRNA | AK122479.1 | - | 5 | 4835 |
Gene Symbol: B230214O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK045604.1 | - | 1 | 35 |
Gene Symbol: BC053393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145729.1 | - | 3 | 1304 |
Gene Symbol: C530040J15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083058.1 | - | 1 | 2085 |
Gene Symbol: Casc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205369.1 | NP_001192298.1 | 10 | 4081 | ||
NM_001205370.1 | NP_001192299.1 | 9 | 3913 | |||
NM_001205371.1 | NP_001192300.1 | 10 | 3671 | |||
NM_177054.5 | NP_796028.2 | 9 | 3982 | |||
NM_199038.3 | NP_950239.2 | 8 | 3814 |
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161213.1 | - | 7 | 2398 |
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC031435.1 | - | 4 | 707 | 4221062 |
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111062.1 | NP_001104532.1 | 6 | 1586 | ||
NM_001111063.1 | NP_001104533.1 | 5 | 1397 | |||
NM_007744.3 | NP_031770.2 | 5 | 1529 | |||
GenBank mRNA | AK133500.1 | - | 1 | 45 | ||
GU324998.1 | - | 1 | 112 |
Gene Symbol: Ctsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028038.1 | - | 828 |
Gene Symbol: Cyp4x1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038526.1 | - | 3290 |
Gene Symbol: D3Ertd254e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101478.1 | NP_001094948.1 | 4 | 2816 | ||
GenBank mRNA | AK153849.1 | - | 4 | 2769 |
Gene Symbol: Dnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC106188.1 | - | 1716 | 30135288 |
Gene Symbol: Efcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159697.1 | NP_001153169.1 | 8 | 2653 |
Gene Symbol: Exoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040212.1 | - | 1 | 517 |
Gene Symbol: Fn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054456.1 | - | 6 | 1988 |
Gene Symbol: Gm3853 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034559.1 | - | 1 | 981 |
Gene Symbol: H2-T23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010398.3 | NP_034528.1 | 7 | 1176 | ||
GenBank mRNA | AK133502.1 | - | 7 | 1176 | ||
BC004093.1 | - | 2296 | ||||
BC005648.1 | - | 1165 | 3709267 | |||
BC099472.1 | - | 5 | 1844 | 30842866 | ||
L00606.1 | - | 1170 | ||||
M11284.1 | - | 1177 |
Gene Symbol: Hook2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161612.1 | - | 16 | 2776 |
Gene Symbol: Kat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK164336.1 | - | 1 | 2884 |
Gene Symbol: LOC100047044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147615.1 | - | 1 | 4260 |
Gene Symbol: Lcmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177846.3 | NP_808514.2 | 1 | 3290 | ||
GenBank mRNA | AK054392.1 | - | 1 | 2980 |
Gene Symbol: Lrrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028838.2 | NP_083114.2 | 9 | 1694 | ||
GenBank mRNA | AJ428068.1 | - | 9 | 1693 |
Gene Symbol: Lyrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175364.4 | NP_780573.1 | 3 | 1211 | ||
GenBank mRNA | AK044791.1 | - | 3 | 1207 | ||
BY465252.1 | - | 1 | 318 |
Gene Symbol: Mfsd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178620.3 | NP_848735.1 | 13 | 2984 | ||
GenBank mRNA | BC088988.1 | - | 13 | 2984 | 6418757 |
Gene Symbol: Mrps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144195.1 | - | 2093 |
Gene Symbol: Ngrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC004034.1 | - | 1216 | 3495639 |
Gene Symbol: Pi4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY148879.1 | - | 2037 | |||
BC010257.1 | - | 1151 |
Gene Symbol: Pola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139486.1 | - | 5 | 571 | ||
AK164285.1 | - | 2 | 3273 |
Gene Symbol: Pola2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164057.1 | NP_001157529.1 | 18 | 2316 | ||
GenBank mRNA | AK087989.1 | - | 18 | 2312 |
Gene Symbol: Polr2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025593.1 | NP_079869.1 | 2 | 1561 | ||
GenBank mRNA | AK149137.1 | - | 2 | 1541 |
Gene Symbol: Ppip5k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC036725.1 | - | 5 | 2083 |
Gene Symbol: Ppm1k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175523.4 | NP_780732.1 | 7 | 3343 | ||
GenBank mRNA | BC092238.1 | - | 7 | 3342 | 30642218 | |
BC138473.1 | - | 6 | 3149 | 8861493 | ||
BC138474.1 | - | 6 | 3147 | 8861494 |
Gene Symbol: Rab33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016858.2 | NP_058554.1 | 2 | 3351 | ||
GenBank mRNA | AB004665.1 | - | 2 | 3318 |
Gene Symbol: Rab42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035722.1 | - | 1 | 2380 |
Gene Symbol: Rad52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M31441.1 | - | 97 |
Gene Symbol: Rin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145495.2 | NP_663470.1 | 10 | 3149 | ||
GenBank mRNA | BC011277.1 | - | 10 | 3149 | 3711198 |
Gene Symbol: Rpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK161487.1 | - | 23 | 4622 | ||
AK164658.1 | - | 23 | 4623 |
Gene Symbol: Rplp2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC055860.1 | - | 1 | 238 |
Gene Symbol: Rundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042981.1 | - | 1 | 36 |
Gene Symbol: Sag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY651760.1 | - | 9 | 1281 |
Gene Symbol: Sif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M11944.1 | - | 1 | 22 |
Gene Symbol: Slc23a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011397.4 | NP_035527.3 | 15 | 2752 | ||
GenBank mRNA | AK143959.1 | - | 15 | 2753 |
Gene Symbol: Srpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK164733.1 | - | 2 | 4811 |
Gene Symbol: St6galnac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC010208.1 | - | 3443 |
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK134171.1 | - | 5 | 2087 |
Gene Symbol: Synrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194341.2 | NP_919322.1 | 20 | 4493 | ||
GenBank mRNA | BC022694.1 | - | 4 | 1538 | ||
BC056370.1 | - | 20 | 4453 | 6831893 |
Gene Symbol: Tmem69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177670.4 | NP_808338.2 | 3 | 1687 |
Gene Symbol: Tmem86b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012462.1 | - | 1478 |
Gene Symbol: Utp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047081.1 | - | 3118 |
Gene Symbol: Wdr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028203.1 | NP_082479.1 | 2 | 2063 | ||
GenBank mRNA | AK011125.1 | - | 3 | 2265 | ||
AK132326.1 | - | 2 | 2035 |
Gene Symbol: Yeats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122463.1 | - | 12 | 5534 |
Gene Symbol: Zfp952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050612.1 | - | 2 | 1118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
