siRNA Details
Gene Symbol: Bpifb9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175167.3 | NP_780376.2 | 3 | 356 | ||
GenBank mRNA | AK029377.1 | - | 3 | 382 | ||
BC047134.1 | - | 3 | 356 | 6308426 | ||
BC080814.1 | - | 3 | 365 | 6391119 |
Gene Symbol: Bpifb9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025574.1 | NP_001020745.1 | 3 | 364 | ||
GenBank mRNA | AK017304.1 | - | 3 | 382 | ||
BC083121.1 | - | 3 | 364 | 6314132 |

More Information
Other Species Targeted

Gene Ontology Categories:
