siRNA Details
Gene Symbol: MELK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256685.1 | NP_001243614.1 | 3 | 264 | ||
NM_001256687.1 | NP_001243616.1 | 3 | 264 | |||
NM_001256688.1 | NP_001243617.1 | 3 | 264 | |||
NM_014791.3 | NP_055606.1 | 3 | 264 | |||
XM_011518076.2 | XP_011516378.1 | 3 | 443 | |||
XM_011518077.1 | XP_011516379.1 | 4 | 339 | |||
XM_011518078.2 | XP_011516380.1 | 4 | 526 | |||
XM_011518079.1 | XP_011516381.1 | 3 | 216 | |||
XM_011518080.1 | XP_011516382.1 | 3 | 231 | |||
XM_011518086.2 | XP_011516388.1 | 3 | 231 | |||
GenBank mRNA | AK222811.1 | - | 3 | 263 | ||
AK293284.1 | - | 3 | 229 | |||
AK299164.1 | - | 3 | 216 | |||
AK300761.1 | - | 3 | 229 | |||
BC014039.2 | - | 3 | 218 | 4547136 | ||
BP365611.1 | - | 3 | 266 | |||
D79997.1 | - | 3 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
