siRNA Details
Gene Symbol: Wdr44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041211.1 | - | 3, 4 | 366 | ||
AK049071.1 | - | 2, 3 | 506 | |||
AK088885.1 | - | 3, 4 | 331 | |||
BB619844.1 | - | 2, 3 | 532 | |||
BC049191.1 | - | 3, 4 | 269 | 6476607 | ||
BC068151.1 | - | 2, 3 | 465 | 30534873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
