siRNA Details
Gene Symbol: Gm3604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162910.1 | NP_001156382.1 | 4 | 300 | ||
GenBank mRNA | BC049144.1 | - | 4 | 292 |
Gene Symbol: 2810408B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013043.1 | - | 729 |
Gene Symbol: 6720489N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173381.1 | NP_775557.1 | 3 | 289 | ||
GenBank mRNA | AK033001.1 | - | 3 | 291 | ||
AK033958.1 | - | 3 | 273 | |||
BC028252.1 | - | 373 | ||||
BC053725.1 | - | 3 | 278 |
Gene Symbol: AU016916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN455334.1 | - | 4 | 303 |
Gene Symbol: Gm5141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256065.1 | NP_001242994.1 | 4 | 333 | ||
GenBank mRNA | BC064714.1 | - | 4 | 333 |
Gene Symbol: Zfp808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039239.2 | NP_001034328.2 | 4 | 344 | ||
GenBank mRNA | BC061203.1 | - | 4 | 344 | 30248220 |
Gene Symbol: Zfp934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162911.1 | NP_001156383.1 | 4 | 318 | ||
NM_175252.4 | NP_780461.2 | 5 | 522 | |||
GenBank mRNA | AK020133.1 | - | 3 | 287 | ||
AK030776.1 | - | 5 | 525 | |||
BC021319.1 | - | 237 | ||||
BC115861.1 | - | 3 | 169 | |||
BC139791.1 | - | 4 | 230 |
Gene Symbol: Zfp935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136496.1 | NP_001129968.1 | 3 | 250 | ||
NM_178875.4 | NP_849206.2 | 4 | 311 | |||
GenBank mRNA | AK018441.1 | - | 3 | 241 | ||
AY189975.1 | - | 3 | 226 | |||
BC012524.1 | - | 3 | 226 | 4008347 | ||
BC037448.1 | - | 3 | 255 | |||
BC047396.1 | - | 4 | 288 | 3979342 | ||
BC087936.1 | - | 4 | 311 | |||
BC113193.1 | - | 4 | 307 | 40062446 | ||
BC115966.1 | - | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
