siRNA Details
Gene Symbol: Cd47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK018679.1 | - | 400 | |||
AK050387.1 | - | 1081 | ||||
AK164165.1 | - | 2832 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
