siRNA Details
Gene Symbol: Atf7ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011885.1 | - | 1 | 217 | ||
AK035245.1 | - | 1 | 2451 | |||
AK050961.1 | - | 1 | 1414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
