siRNA Details
Gene Symbol: NUAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014840.2 | NP_055655.1 | 7 | 2414 | ||
GenBank mRNA | AB011109.1 | - | 7 | 2414 | ||
AK289992.1 | - | 7 | 1094 | |||
BC152462.1 | - | 7 | 2414 | 8862598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
