siRNA Details
Gene Symbol: 1700055D18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006794.1 | - | 1 | 448 | ||
AK042394.1 | - | 1 | 1311 | |||
AV204476.1 | - | 1 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
