siRNA Details
Gene Symbol: C130026I21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037909.3 | NP_001032998.2 | 7 | 1586 | ||
NM_175219.3 | NP_780428.1 | 8 | 1670 | |||
GenBank mRNA | AK047983.1 | - | 8 | 1648 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
