siRNA Details
Gene Symbol: Zfp934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162911.1 | NP_001156383.1 | 4 | 1500 | ||
NM_175252.4 | NP_780461.2 | 6 | 1368 | |||
GenBank mRNA | AK030776.1 | - | 6 | 1371 | ||
BC021319.1 | - | 1419 | ||||
BC115861.1 | - | 3 | 1351 |
