siRNA Details
Gene Symbol: Nav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111016.1 | NP_001104486.1 | 6 | 1023 | ||
NM_175272.3 | NP_780481.2 | 5 | 1285 | |||
GenBank mRNA | AK031334.1 | - | 5 | 583 | ||
AK039374.1 | - | 5 | 1293 | |||
AK040152.1 | - | 4 | 570 | |||
AK084063.1 | - | 6 | 1001 | |||
AK129480.1 | - | 4 | 511 | |||
AK158337.1 | - | 5 | 697 | |||
BC150679.1 | - | 4 | 273 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_138529, XM_008759314, XM_006229244, XM_008759312, XM_008759313.
Gene Ontology Categories:
