siRNA Details
Gene Symbol: Dock10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285927.1 | NP_001272856.1 | 48 | 5514 | ||
NM_175291.4 | NP_780500.2 | 48 | 5498 | |||
GenBank mRNA | AK034064.1 | - | 3 | 229 | ||
AK122347.1 | - | 38 | 4253 | |||
BC150753.1 | - | 48 | 5434 | 9087664 | ||
CX224190.1 | - | 4 | 450 | |||
GU797481.1 | - | 48 | 5471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
