siRNA Details
Gene Symbol: Sall4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049188.1 | - | 2 | 591 | ||
AK145388.1 | - | 2 | 609 | |||
AY463371.1 | - | 2 | 491 | |||
AY463373.1 | - | 2 | 491 | |||
BC053716.1 | - | 2 | 659 | |||
BC099928.1 | - | 2 | 614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
