siRNA Details
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030732.3 | NP_109657.2 | 16 | 5787 | ||
GenBank mRNA | AK082694.1 | - | 1 | 600 | ||
AK140144.1 | - | 1 | 1556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
