siRNA Details
Gene Symbol: Adamts9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175314.3 | NP_780523.2 | 15 | 2058 | ||
GenBank mRNA | AK030699.1 | - | 2 | 168 | ||
AK173146.1 | - | 2 | 178 | |||
BC068142.1 | - | 1 | 97 | |||
CK793536.1 | - | 5 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
