siRNA Details
Gene Symbol: Cphx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175342.3 | NP_780551.1 | 4 | 447 | ||
GenBank mRNA | AK049114.1 | - | 448 | |||
BC095973.1 | - | 419 | ||||
BC132038.1 | - | 445 | 40130315 | |||
BC132040.1 | - | 445 | 40130317 | |||
DQ224405.1 | - | 499 |
Gene Symbol: Cphx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270506.1 | NP_001257435.1 | 4 | 451 | ||
GenBank mRNA | DV647390.1 | - | 4 | 434 |
