siRNA Details
Gene Symbol: Trps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032000.2 | NP_114389.2 | 6 | 6415 | ||
GenBank mRNA | AK046585.1 | - | 1 | 922 | ||
AK142355.1 | - | 6 | 6084 | |||
BC083110.1 | - | 5 | 5174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
