siRNA Details
Gene Symbol: Ibtk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081282.2 | NP_001074751.1 | 26 | 4178 | ||
GenBank mRNA | AK031796.1 | - | 15 | 1979 | ||
AK077387.1 | - | 4 | 321 | |||
AK129353.1 | - | 23 | 3056 | |||
BC050255.1 | - | 14 | 3612 | |||
BC060722.1 | - | 25 | 3972 | |||
BC137798.1 | - | 26 | 4067 | 8860817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
