siRNA Details
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175375.3 | NP_780584.2 | 29 | 6392 | ||
GenBank mRNA | AK078916.1 | - | 1 | 359 | ||
AK129284.1 | - | 629 | ||||
BC091647.1 | - | 5 | 1817 |
Gene Symbol: Maskbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC016604.1 | - | 1 | 294 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
