siRNA Details
Gene Symbol: Dock10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285927.1 | NP_001272856.1 | 50 | 5724 | ||
NM_175291.4 | NP_780500.2 | 50 | 5708 | |||
GenBank mRNA | AK034064.1 | - | 5 | 439 | ||
AK122347.1 | - | 40 | 4463 | |||
BC150753.1 | - | 50 | 5644 | 9087664 | ||
GU797481.1 | - | 50 | 5681 |

More Information
Other Species Targeted

Gene Ontology Categories:
