siRNA Details
Gene Symbol: Plekhg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081333.1 | NP_001074802.1 | 12, 13 | 2182 | ||
GenBank mRNA | AK029859.1 | - | 10, 11 | 2854 |

More Information
Other Species Targeted

Gene Ontology Categories:
