siRNA Details
Gene Symbol: Maskbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003441.1 | - | 1 | 35 | ||
BC016604.1 | - | 1 | 460 |
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175375.3 | NP_780584.2 | 29 | 6558 | ||
GenBank mRNA | AK078916.1 | - | 1 | 525 | ||
AK129284.1 | - | 795 | ||||
BC091647.1 | - | 5 | 1983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
