siRNA Details
Gene Symbol: Trim9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043347.1 | - | 1 | 3235 | ||
AK047483.1 | - | 1 | 703 | |||
AK136860.1 | - | 1 | 1291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
