siRNA Details
Gene Symbol: Cntln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175275.4 | NP_780484.2 | 16 | 2868 | ||
GenBank mRNA | AK029118.1 | - | 2 | 397 | ||
BC089374.1 | - | 2 | 571 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
