siRNA Details
Gene Symbol: A430005L14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163019.1 | NP_001156491.1 | 3 | 809 | ||
NM_175287.4 | NP_780496.2 | 4 | 923 | |||
GenBank mRNA | AK039812.1 | - | 4 | 924 | ||
AK207640.1 | - | 2 | 122 | |||
BC052148.1 | - | 4 | 923 | 6706797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
