siRNA Details
Gene Symbol: Nlrp4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175290.4 | NP_780499.3 | 7 | 2579 | ||
GenBank mRNA | AK049352.1 | - | 5 | 982 | ||
BC098479.1 | - | 5 | 1001 |
Gene Symbol: Nlrp4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY596195.1 | - | 8 | 2361 |
Gene Symbol: Nlrp4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136247.1 | - | 3049 |

More Information
Other Species Targeted

Gene Ontology Categories:
